Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fs(1)h
Entrez Gene ID
31722
Gene Name
female sterile (1) homeotic
Gene Aliases
Brd4, CG2252, Dmel\CG2252, Dmel_CG2252, FSH, Fs(1)h, Fs(1)h-L, Fs(1)h/Brd4, Fs(1)hL, Fs(1)hS, Fsh, N72, ORE-1, anon-EST:Liang-1.81, anon-EST:fe1G2, clone 1.81, fs(1)1456, fs(1)26/26A, fs(1)M16, fs(1)R10.4, fsh, fst(1)h, l(1)7Da, l(1)G0093, l(1)G0495, rnc
Chromosome Location
Chr.X: 8037808 - 8061436 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004656
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272392.2 NP_001259321.1 4 - 5 3701 69
NM_078523.3 NP_511078.2 4 - 5 3701 69

View Full Product Details