Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hk
Entrez Gene ID
31955
Gene Name
Hyperkinetic
Gene Aliases
CG17779, CG2287, CG32688, CG34319, CG43388, Dm_X:51771, Dmel\CG43388, Dmel_CG43388, hk, lincRNA.975
Chromosome Location
Chr.X: 10233347 - 10263485 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0263220
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258680.2 NP_001245609.1 11 - 12 2526 66
NM_001258681.2 NP_001245610.1 8 - 9 2268 66
NM_001258682.2 NP_001245611.1 10 - 11 2395 66
NM_001258683.2 NP_001245612.1 10 - 11 2016 66
NM_001272470.2 NP_001259399.1 5 - 6 2031 66
NM_001272471.2 NP_001259400.1 5 - 6 2028 66
NM_001272472.2 NP_001259401.1 5 - 6 2028 66
NM_078549.4 NP_511104.3 5 - 6 2028 66
NM_167222.3 NP_727405.1 10 - 11 2013 66
GenBank mRNA AY060359.1 - 5 - 6 1157 66

View Full Product Details