Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ten-a
Entrez Gene ID
32183
Gene Name
Tenascin accessory
Gene Aliases
1.2, CG11270, CG12720, CG15733, CG18182, CG2578, CG2590, CG32659, CG42338, Dmel\CG42338, Dmel_CG42338, DmtenA, Dmtena, Ten11A, Ten[a], cbd, ten-a, tenA, tenA(AT)[[14]], ten[a]
Chromosome Location
Chr.X: 12043698 - 12335214 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0267001
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144717.4 NP_001138189.1 10 - 11 4236 82
NM_001144718.3 NP_001138190.1 11 - 12 4941 82
NM_001169258.2 NP_001162729.1 5 - 6 2170 82
NM_001169259.3 NP_001162730.2 11 - 12 4346 82
NM_001169260.3 NP_001162731.1 12 - 13 5212 82
NM_001169261.3 NP_001162732.2 10 - 11 4236 82
NM_001258711.2 NP_001245640.1 8 - 9 3185 82
NM_001258712.2 NP_001245641.1 8 - 9 3191 82
NM_001258713.2 NP_001245642.1 8 - 9 3203 82
NM_001272554.2 NP_001259483.1 8 - 9 3085 82
NM_078582.4 NP_511137.3 6 - 7 2371 82

View Full Product Details