Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NetB
Entrez Gene ID
32400
Gene Name
Netrin-B
Gene Aliases
CG10521, CG15021, CT29512, Dmel\CG10521, Dmel_CG10521, NETB, NetrinB, net, netB, netrin, netrin B
Chromosome Location
Chr.X: 14686038 - 14749376 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015774
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169282.1 NP_001162753.1 7 - 8 2627 66
NM_001169283.2 NP_001162754.1 8 - 9 2630 66
NM_001169284.2 NP_001162755.1 8 - 9 2567 66
NM_001258739.2 NP_001245668.1 8 - 9 2633 66
NM_001258740.2 NP_001245669.1 8 - 9 2570 66
NM_001272617.1 NP_001259546.1 7 - 8 2606 66
NM_078600.4 NP_511155.1 8 - 9 2720 66
GenBank mRNA U60317.1 - 8 - 9 2617 66
U63737.1 - 7 - 8 2566 66

View Full Product Details