Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
baz
Entrez Gene ID
32703
Gene Name
bazooka
Gene Aliases
BAZ, Baz, Baz/Par-3, Baz/Par3, Bazooka, CG5055, D-Par3, D-par3, Dmel\CG5055, Dmel_CG5055, PAR-3, PAR3, Par-3, Par3, dPar-3, l(1)G0484, par-3, par3
Chromosome Location
Chr.X: 17160006 - 17200728 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000163
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298435.2 NP_001285364.1 1 - 2 1185 75
NM_001347807.1 NP_001334669.1 1 - 2 1185 75
GenBank mRNA BT032834.1 - 1 - 2 256 75

View Full Product Details