Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
baz
Entrez Gene ID
32703
Gene Name
bazooka
Gene Aliases
BAZ, Baz, Baz/Par-3, Baz/Par3, Bazooka, CG5055, D-Par3, D-par3, Dmel\CG5055, Dmel_CG5055, PAR-3, PAR3, Par-3, Par3, dPar-3, l(1)G0484, par-3, par3
Chromosome Location
Chr.X: 17160006 - 17200728 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000163
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042818.4 NP_001036283.1 4 - 5 6838 93
NM_001298435.2 NP_001285364.1 2 - 3 2884 93
NM_001298436.2 NP_001285365.1 4 - 5 6838 93
NM_001347807.1 NP_001334669.1 2 - 3 2884 93
GenBank mRNA BT032834.1 - 2 - 3 1955 93

View Full Product Details