Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- OdsH
- Entrez Gene ID
-
32758
- Gene Name
- Ods-site homeobox
- Gene Aliases
-
CG6352, Dmel\CG6352, Dmel_CG6352, OdsH[mel], Odsh, odsH, odsh
- Chromosome Location
-
Chr.X: 17791010 - 17814011 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026058
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078665.3 |
NP_523389.3 |
2 - 3 |
662 |
62 |
GenBank mRNA |
BT022139.1 |
- |
2 - 3 |
662 |
62 |
View Full Product Details