Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Bx
- Entrez Gene ID
-
32846
- Gene Name
- Beadex
- Gene Aliases
-
Bd, CG10548, CG15048, CG44425, CG6500, DLMO, Dlmo, Dmel\CG44425, Dmel_CG44425, LMO, Lmo, Ptd, Rho, beadex/dLMO, bx, dLMO, dLmo, dlmo, dttg, fliH, hdp, hdp-a, hld
- Chromosome Location
-
Chr.X: 18515428 - 18572809 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0265598
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078678.4 |
NP_523402.4 |
1 - 2 |
315 |
76 |
GenBank mRNA |
BT004844.1 |
- |
1 - 2 |
284 |
76 |
View Full Product Details