Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nep3
Entrez Gene ID
33005
Gene Name
Neprilysin 3
Gene Aliases
CEC, CG9565, DmeNEP3, Dmel\CG9565, Dmel_CG9565, NEP3, dNEP3, nep3
Chromosome Location
Chr.X: 19961297 - 19969323 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031081
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258846.1 NP_001245775.1 2 - 3 283 57
NM_001258847.2 NP_001245776.1 2 - 3 421 57
NM_078693.3 NP_523417.2 2 - 3 274 57
GenBank mRNA BT132915.1 - 2 - 3 274 57

View Full Product Details