Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ser6
Entrez Gene ID
33073
Gene Name
Serine protease 6
Gene Aliases
CG2071, Dmel\CG2071, Dmel_CG2071, SP47
Chromosome Location
Chr.X: 20962605 - 20963505 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011834
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078702.2 NP_523426.1 1 - 1 746 97
GenBank mRNA AY089642.1 - 1 - 1 743 97

View Full Product Details