Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or23a
- Entrez Gene ID
-
33450
- Gene Name
- Odorant receptor 23a
- Gene Aliases
-
23A.1, 23a, AN5, CG9880, DOR23A.1, DOR64, Dmel\CG9880, Dmel_CG9880, Dor64, Or23A.1, Or64, dor64, or23a
- Chromosome Location
-
Chr.2L: 2654049 - 2655462 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026395
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078734.4 |
NP_523458.3 |
1 - 2 |
808 |
62 |
GenBank mRNA |
AF127925.1 |
- |
1 - 2 |
735 |
62 |
View Full Product Details