Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gart
Entrez Gene ID
33986
Gene Name
GART trifunctional enzyme
Gene Aliases
143062_at, ADE8, Ade3, CG31628, CG8761, D, Dmel\CG31628, Dmel_CG31628, GART, Gart/ade3, ade3, gart, l(2)27Da
Chromosome Location
Chr.2L: 7014623 - 7023898 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000053
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298769.1 NP_001285698.1 6 - 7 2964 129
NM_078773.3 NP_523497.2 6 - 7 2964 129

View Full Product Details