Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pvf2
Entrez Gene ID
33994
Gene Name
PDGF- and VEGF-related factor 2
Gene Aliases
CG13780, DmVEGF-2, Dmel\CG13780, Dmel_CG13780, PV2, PVF, PVF2, PVFs, Pvf, VEGF, VEGF-2, VEGF27Cb, Vegf27Cb, pvf2
Chromosome Location
Chr.2L: 7069536 - 7084635 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031888
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078775.3 NP_523499.2 3 - 4 806 87
GenBank mRNA AJ312312.1 - 2 - 3 611 87
AY084196.1 - 3 - 4 807 87

View Full Product Details