Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kua
Entrez Gene ID
35300
Gene Name
CG10723-PA
Gene Aliases
38B.18, CG10723, Dmel\CG10723, Dmel_CG10723
Chromosome Location
Chr.2L: 20097581 - 20103375 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032850
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201927.2 NP_001188856.1 2 - 3 624 66
NM_001273691.1 NP_001260620.1 2 - 3 347 66
NM_001273692.1 NP_001260621.1 3 - 4 835 66
NM_136182.2 NP_610026.1 2 - 3 328 66
GenBank mRNA AF152361.1 - 1 - 2 242 66
BT029029.1 - 2 - 3 328 66
BT126266.1 - 3 - 4 835 66

View Full Product Details