Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pwn
Entrez Gene ID
44011
Gene Name
pawn
Gene Aliases
CG11101, CT31067, Dmel\CG11101, Dmel_CG11101, l(2)pwn
Chromosome Location
Chr.2R: 7282572 - 7300806 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003174
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273834.1 NP_001260763.1 9 - 10 4207 82

View Full Product Details