Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cse1
- Entrez Gene ID
-
35016
- Gene Name
- Chromosome segregation 1
- Gene Aliases
-
BcDNA:LD14270, CAS, CG13281, Cas, Dcas, Dmel\CG13281, Dmel_CG13281, cas, dcas, l(2)k03902
- Chromosome Location
-
Chr.2L: 16821156 - 16825207 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0022213
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078864.3 |
NP_523588.2 |
6 - 7 |
2330 |
111 |
View Full Product Details