Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Tsp42Ej
- Entrez Gene ID
-
35620
- Gene Name
- Tetraspanin 42Ej
- Gene Aliases
-
CG12143, Dm.Tsp42Ej, Dmel\CG12143, Dmel_CG12143, Sun, sun, tsp42ej
- Chromosome Location
-
Chr.2R: 7040136 - 7042762 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033132
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078912.3 |
NP_523636.1 |
1 - 2 |
405 |
60 |
GenBank mRNA |
AY058262.1 |
- |
1 - 2 |
312 |
60 |
View Full Product Details