Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tsp42Ek
Entrez Gene ID
35621
Gene Name
Tetraspanin 42Ek
Gene Aliases
CG12841, Dm.Tsp42Ek, Dmel\CG12841, Dmel_CG12841
Chromosome Location
Chr.2R: 7042656 - 7044710 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033133
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259232.1 NP_001246161.1 1 - 2 434 83
NM_078913.4 NP_523637.1 1 - 2 180 83
GenBank mRNA BT088928.1 - 1 - 2 434 83

View Full Product Details