Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Tsp42Eo
- Entrez Gene ID
-
35625
- Gene Name
- Tetraspanin 42Eo
- Gene Aliases
-
BcDNA:RH51444, CG12838, Dm.Tsp42Eo, Dmel\CG12838, Dmel_CG12838
- Chromosome Location
-
Chr.2R: 7054717 - 7056576 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033136
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078917.3 |
NP_523641.2 |
1 - 2 |
223 |
90 |
GenBank mRNA |
AY070986.1 |
- |
1 - 2 |
219 |
90 |
View Full Product Details