Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gr43a
Entrez Gene ID
35655
Gene Name
Gustatory receptor 43a
Gene Aliases
43C.1, 43C1, CG1712, DmGr43a, Dmel\CG1712, Dmel_CG1712, GR43C.1, GR43a, Gr43C1, gr43a
Chromosome Location
Chr.2R: 7390869 - 7393161 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041243
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043066.1 NP_001036531.1 2 - 3 220 71
NM_078926.3 NP_523650.2 2 - 3 220 71
GenBank mRNA BT023089.1 - 2 - 3 220 71

View Full Product Details