Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myd88
Entrez Gene ID
35956
Gene Name
CG2078-PA
Gene Aliases
CG2078, DMMYD88, DmMyD88, DmMyd88, Dmel\CG2078, Dmel_CG2078, EP(2)2535, Kra, LD20892, MYD88, MyD88, Myd88F, dMYd88, dMyD88, dMyd88, kra, myd88
Chromosome Location
Chr.2R: 9302823 - 9308722 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033402
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273892.1 NP_001260821.1 3 - 4 1910 97
NM_001299317.1 NP_001286246.1 3 - 4 1576 97
NM_136635.3 NP_610479.1 3 - 4 1910 97
GenBank mRNA AF422803.1 - 3 - 4 1553 97
AY069468.1 - 3 - 4 1855 97

View Full Product Details