Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pof
Entrez Gene ID
37947
Gene Name
Painting of fourth
Gene Aliases
CG3691, Dmel\CG3691, Dmel_CG3691, POF, cg3691, pof
Chromosome Location
Chr.2R: 24669397 - 24671577 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035047
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259569.2 NP_001246498.1 2 - 3 403 79
NM_079127.3 NP_523851.1 2 - 3 407 79
NM_166682.3 NP_726472.1 1 - 2 456 79
GenBank mRNA AJ295236.1 - 1 - 2 384 79
AY052060.1 - 1 - 2 363 79

View Full Product Details