Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zip
Entrez Gene ID
38001
Gene Name
zipper
Gene Aliases
CG15792, DROMHC, Dm nmII, Dmel\CG15792, Dmel_CG15792, DmnmII, Dronm-MII, E(br), MHC, Mhc-c, Mhc-c[1], Myo, Myo II, Myo-II, Myo-II HC, Myo15, Myo2, MyoII, NMM, NMMII, ZIP, Zip, anon-WO0140519.37, l(2)02957, l(2)17F1, myo II, myoII, myosin-II, nmMHC, nmy-2, zip/MyoII, zpr
Chromosome Location
Chr.2R: 24990570 - 25011965 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0265434
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079136.4 NP_523860.2 5 - 6 886 89
NM_166704.4 NP_726506.1 5 - 6 822 89

View Full Product Details