Assay Design
The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Gene Transcripts
- Gene Symbol
- pan
- Entrez Gene ID
-
43769
- Gene Name
- pangolin
- Gene Aliases
-
CG17964, CG32005, CG34403, DTCF, DTcf, Dm Pan, Dmel\CG34403, Dmel_CG34403, IA5, LEF, LEF-1, LEF/TCF, LEF/TCF-1, LEF1, LEF1/TCF, Lef, Lef1, PAN, Pan, TCF, TCF/LEF, TCF/LEF1, TCF/Pan, Tcf, Tcf-1, Tcf/LEF, d-TCF, dTCF, dTcf, l(4)102ABb, l(4)13, lef1, pan.dTCF, tcf
- Chromosome Location
-
Chr.4: 69326 - 114270 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0085432
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166724.4 |
NP_726528.2 |
1 - 1 |
2844 |
118 |
View Full Product Details