Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ephrin
Entrez Gene ID
43799
Gene Name
CG1862-PA
Gene Aliases
CG1862, Dmel\CG1862, Dmel_CG1862, ephrin, lincRNA.922
Chromosome Location
Chr.4: 571755 - 580122 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040324
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_143670.5 NP_651927.1 2 - 3 917 124
NM_166770.2 NP_726583.1 2 - 3 1020 124
NM_166771.2 NP_726584.1 2 - 3 1225 124
GenBank mRNA AF216287.1 - 2 - 3 865 124
BT005199.1 - 2 - 3 981 124

View Full Product Details