Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fidipidine
Entrez Gene ID
36808
Gene Name
CG7773-PA
Gene Aliases
CG7773, Dmel\CG7773, Dmel_CG7773
Chromosome Location
Chr.2R: 16262284 - 16264619 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025519
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079036.5 NP_523760.1 3 - 4 545 79
GenBank mRNA BT031141.1 - 3 - 4 477 79

View Full Product Details