Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DAT
Entrez Gene ID
36849
Gene Name
Dopamine transporter
Gene Aliases
CG8380, Dat, DmDAT, Dmel\CG8380, Dmel_CG8380, Fumin, dDAT, dat, fmn, fumin
Chromosome Location
Chr.2R: 16558567 - 16565238 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034136
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274097.2 NP_001261026.1 3 - 4 643 81
NM_079039.4 NP_523763.2 3 - 4 643 81
GenBank mRNA AF439752.1 - 2 - 3 376 81
AY051579.1 - 3 - 4 663 81

View Full Product Details