Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- gek
- Entrez Gene ID
-
37858
- Gene Name
- genghis khan
- Gene Aliases
-
CG4012, Dmel\CG4012, Dmel_CG4012, Gek, MRCK, MRCKalpha, MdaPk, Mrck, anon-WO0200864.1, dMRCK, l(2)09373
- Chromosome Location
-
Chr.2R: 24092079 - 24098636 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0023081
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079113.3 |
NP_523837.2 |
2 - 3 |
359 |
121 |
GenBank mRNA |
AY051698.1 |
- |
2 - 3 |
340 |
121 |
View Full Product Details