Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gek
Entrez Gene ID
37858
Gene Name
genghis khan
Gene Aliases
CG4012, Dmel\CG4012, Dmel_CG4012, Gek, MRCK, MRCKalpha, MdaPk, Mrck, anon-WO0200864.1, dMRCK, l(2)09373
Chromosome Location
Chr.2R: 24092079 - 24098636 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0023081
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079113.3 NP_523837.2 6 - 7 4243 83
GenBank mRNA AY051698.1 - 6 - 7 4224 83

View Full Product Details