Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
key
Entrez Gene ID
37967
Gene Name
kenny
Gene Aliases
CG16910, DmIKK-gamma, DmIKKgamma, Dmel\CG16910, Dmel_CG16910, Dmikkgamma, IKK, IKK-gamma, IKKG, IKK[[gamma]], IKKg, IKKgamma, Ikkgammabeta, KEY, Kenny, Key, dIKK, dIKK-gamma, dIKKgamma, dmIKKgamma
Chromosome Location
Chr.2R: 24785514 - 24787411 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041205
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259574.2 NP_001246503.1 4 - 5 891 66
NM_079132.4 NP_523856.2 4 - 5 891 66

View Full Product Details