Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FoxL1
Entrez Gene ID
38471
Gene Name
Forkhead box L1
Gene Aliases
CG1132, Dmel\CG1132, Dmel_CG1132, Dmfd2, FD2, Fd64A, fd2, fd64A, foxL1
Chromosome Location
Chr.3L: 4018878 - 4021279 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004895
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259680.2 NP_001246609.1 1 - 2 585 66
NM_079188.3 NP_523912.1 1 - 2 543 66
GenBank mRNA BT120383.1 - 1 - 2 369 66

View Full Product Details