Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- nAChRbeta1
- Entrez Gene ID
-
38545
- Gene Name
- nicotinic Acetylcholine Receptor beta1
- Gene Aliases
-
AChR, AChR64B, ARD, AchR64B, Acr64B, AcrD, Ard, CG11348, CG11348-PA, CG12606, Dbeta1, Dmel\CG11348, Dmel_CG11348, ard, beta 64B, beta1 nAChR, nAChR, nAChR-beta64B, nAcR64B, nAcRbeta-64B
- Chromosome Location
-
Chr.3L: 4429349 - 4435993 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000038
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079203.3 |
NP_523927.2 |
6 - 7 |
1645 |
58 |
GenBank mRNA |
BT125991.1 |
- |
6 - 7 |
1623 |
58 |
View Full Product Details