Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Arr2
- Entrez Gene ID
-
38993
- Gene Name
- Arrestin 2
- Gene Aliases
-
49, AH, ARR2, Arr, Arr-2, ArrB, ArrB-ml, CG5962, DAII, DmArrestin-2, Dmel\CG5962, Dmel_CG5962, Dro.-Arr.II, PRI, arr, arr2
- Chromosome Location
-
Chr.3L: 8647261 - 8648936 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000121
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079252.3 |
NP_523976.1 |
2 - 3 |
1160 |
59 |
GenBank mRNA |
BT031017.1 |
- |
2 - 3 |
1156 |
59 |
View Full Product Details