Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fdx1
Entrez Gene ID
39070
Gene Name
Ferredoxin 1
Gene Aliases
BcDNA:RH67819, CG4205, Dmel\CG4205, Dmel_CG4205, Fdxh
Chromosome Location
Chr.3L: 9370195 - 9371238 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011769
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202146.1 NP_001189075.1 2 - 3 166 73
NM_079269.3 NP_523993.1 1 - 2 158 73

View Full Product Details