Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CalpB
Entrez Gene ID
39165
Gene Name
Calpain-B
Gene Aliases
CALPB, CG8107, Dmel\CG8107, Dmel_CG8107, calpB
Chromosome Location
Chr.3L: 9888762 - 9893319 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025866
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259777.2 NP_001246706.1 2 - 3 1397 74
NM_079292.5 NP_524016.4 2 - 3 1739 74
GenBank mRNA BT031259.1 - 2 - 3 1386 74

View Full Product Details