Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pi3K68D
Entrez Gene ID
39329
Gene Name
Phosphatidylinositol 3-kinase 68D
Gene Aliases
11621, BcDNA:LD15217, CG11621, Cpk, Dmel\CG11621, Dmel_CG11621, PI(3)K, PI3K, PI3K 68D, PI3K 68_D, PI3K-68D, PI3K-68D/E, PI3K68D, PI3K_68D, PI[[3]]K, Pi3K, cpk, dPI3K, dPI3k, dPIK, pi3k68d
Chromosome Location
Chr.3L: 11774650 - 11790013 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015278
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169949.2 NP_001163420.1 12 - 13 4972 71
NM_079304.4 NP_524028.2 11 - 12 5321 71
NM_168475.3 NP_729743.1 12 - 13 5020 71
GenBank mRNA AY058587.1 - 11 - 12 5263 71

View Full Product Details