Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Drl-2
Entrez Gene ID
36436
Gene Name
Derailed 2
Gene Aliases
CG12463, CG3915, DNT-like, DRL-2, Dmel\CG3915, Dmel_CG3915, dnl, drl-2, drl2
Chromosome Location
Chr.2R: 13055246 - 13079539 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033791
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299442.1 NP_001286371.1 2 - 3 1333 95
NM_136991.4 NP_610835.2 2 - 3 1333 95
GenBank mRNA BT004493.1 - 2 - 3 1304 95

View Full Product Details