Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prosap
Entrez Gene ID
50225
Gene Name
CG30483-PA
Gene Aliases
BcDNA:GH23107, CG13354, CG30483, CG8122, D-ProSAP, Dmel\CG30483, Dmel_CG30483, ProSAP, Shank, anon-WO0118547.149, prosap
Chromosome Location
Chr.2R: 14060456 - 14140902 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040752
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259386.2 NP_001246315.2 8 - 9 2835 66
NM_137081.5 NP_610925.3 9 - 10 3030 66
GenBank mRNA BT044467.1 - 9 - 10 2113 66

View Full Product Details