Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zasp52
Entrez Gene ID
36740
Gene Name
Z band alternatively spliced PDZ-motif protein 52
Gene Aliases
CG12968, CG12969, CG30084, CG8242, Dmel\CG30084, Dmel_CG30084, Z(210), ZASP52, Zasp, Zbp210, dzasp, tun, tungus, zasp, zasp52
Chromosome Location
Chr.2R: 15762565 - 15817004 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0265991
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137214.5 NP_611058.4 9 - 10 1376 66
NM_145757.3 NP_665700.2 9 - 10 1432 66

View Full Product Details