Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tun
Entrez Gene ID
36743
Gene Name
tungus
Gene Aliases
BcDNA:LD38553, CG8253, Dmel\CG8253, Dmel_CG8253, Ntaq1
Chromosome Location
Chr.2R: 15794000 - 15855645 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034046
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259422.2 NP_001246351.1 1 - 2 1102 83
NM_001259423.2 NP_001246352.1 1 - 2 1102 83
NM_001299539.1 NP_001286468.1 2 - 3 2122 83
NM_137217.3 NP_611061.1 1 - 2 1102 83
GenBank mRNA AY060439.1 - 1 - 2 1075 83

View Full Product Details