Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rg
Entrez Gene ID
44531
Gene Name
rugose
Gene Aliases
AKAP, Akap550, CG15464, CG15466, CG44835, CG6775, DAKAP550, Dmel\CG44835, Dmel_CG44835, fas
Chromosome Location
Chr.X: 5085759 - 5254864 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266098
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042796.3 NP_001036261.3 17 - 18 3614 72
NM_001144685.2 NP_001138157.2 17 - 18 3926 72
NM_001144686.2 NP_001138158.2 17 - 18 3956 72
NM_001144687.2 NP_001138159.2 16 - 17 3335 72
NM_001297949.1 NP_001284878.1 20 - 21 7943 72
NM_001297950.1 NP_001284879.1 17 - 18 3956 72
NM_167028.3 NP_726978.2 17 - 18 4666 72

Gene Symbol
CG15465
Entrez Gene ID
31434
Gene Name
CG15465-PA
Gene Aliases
Dmel\CG15465, Dmel_CG15465
Chromosome Location
Chr.X: 5085759 - 5254864 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029746
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297948.1 NP_001284877.1 20 - 21 7943 72
NM_131978.3 NP_572206.1 17 - 18 4666 72

View Full Product Details