Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rg
Entrez Gene ID
44531
Gene Name
rugose
Gene Aliases
AKAP, Akap550, CG15464, CG15466, CG44835, CG6775, DAKAP550, Dmel\CG44835, Dmel_CG44835, fas
Chromosome Location
Chr.X: 5085759 - 5254864 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266098
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042796.3 NP_001036261.3 21 - 22 6960 64
NM_001144685.2 NP_001138157.2 21 - 22 7272 64
NM_001144686.2 NP_001138158.2 21 - 22 7302 64
NM_001144687.2 NP_001138159.2 20 - 21 6681 64
NM_001297949.1 NP_001284878.1 24 - 25 11289 64
NM_001297950.1 NP_001284879.1 21 - 22 7302 64
NM_167028.3 NP_726978.2 21 - 22 8012 64

Gene Symbol
CG15465
Entrez Gene ID
31434
Gene Name
CG15465-PA
Gene Aliases
Dmel\CG15465, Dmel_CG15465
Chromosome Location
Chr.X: 5085759 - 5254864 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029746
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297948.1 NP_001284877.1 24 - 25 11289 64
NM_131978.3 NP_572206.1 21 - 22 8012 64

View Full Product Details