Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LysRS
Entrez Gene ID
31904
Gene Name
Lysyl-tRNA synthetase
Gene Aliases
Aats-lys, CG12141, Dmel\CG12141, Dmel_CG12141, KRS, dKARS
Chromosome Location
Chr.X: 9606363 - 9608884 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027084
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201642.2 NP_001188571.1 4 - 5 1973 68
NM_132345.2 NP_572573.1 3 - 4 1782 68
NM_167196.3 NP_727353.1 3 - 4 2036 68
GenBank mRNA AY089547.1 - 3 - 4 1695 68
BT133498.1 - 3 - 4 1794 68

View Full Product Details