Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gr9a
- Entrez Gene ID
-
318158
- Gene Name
- Gustatory receptor 9a
- Gene Aliases
-
CG32693, Dmel\CG32693, Dmel_CG32693, GR9a
- Chromosome Location
-
Chr.X: 10061627 - 10062785 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0052693
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_167217.1 |
NP_727392.1 |
1 - 2 |
812 |
64 |
View Full Product Details