Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
X11Lbeta
Entrez Gene ID
31997
Gene Name
CG32677-PA
Gene Aliases
CG1852, CG1861, CG32677, Dmel\CG32677, Dmel_CG32677, Dmint2, X11B, X11LB, X11lbeta, x11Lbeta
Chromosome Location
Chr.X: 10627737 - 10710947 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0052677
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272496.2 NP_001259425.2 6 - 7 5030 55
NM_001298156.1 NP_001285085.1 8 - 9 4157 55
NM_167239.4 NP_727440.3 8 - 9 4112 55

View Full Product Details