Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eIF4G1
Entrez Gene ID
43839
Gene Name
eukaryotic translation initiation factor 4G1
Gene Aliases
CG10811, Dmel\CG10811, Dmel_CG10811, EIF4G, Eif4G, deIF4G, eIF-4G, eIF4G, eIF4g, p200
Chromosome Location
Chr.4: 915297 - 930715 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0023213
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103382.2 NP_001096852.1 12 - 13 5380 116
NM_001144385.2 NP_001137857.1 11 - 12 4851 116
NM_079901.4 NP_524640.3 11 - 12 4621 116
GenBank mRNA AF030155.1 - 11 - 12 4627 116
BT053671.1 - 11 - 12 4615 116

View Full Product Details