Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- lbk
- Entrez Gene ID
-
36788
- Gene Name
- lambik
- Gene Aliases
-
CG8434, Dmel\CG8434, Dmel_CG8434, Lbk
- Chromosome Location
-
Chr.2R: 16150123 - 16169519 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034083
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137247.3 |
NP_611091.2 |
6 - 7 |
3218 |
65 |
View Full Product Details