Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- APC10
- Entrez Gene ID
-
36975
- Gene Name
- Anaphase Promoting Complex subunit 10
- Gene Aliases
-
Apc10, CG11419, Dmel\CG11419, Dmel_CG11419
- Chromosome Location
-
Chr.2R: 17517261 - 17518186 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034231
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137379.5 |
NP_611223.4 |
1 - 2 |
217 |
65 |
GenBank mRNA |
AY071207.2 |
- |
1 - 2 |
214 |
65 |
View Full Product Details