Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UQCR-6.4
Entrez Gene ID
36991
Gene Name
Ubiquinol-cytochrome c reductase 6.4 kDa subunit
Gene Aliases
CG14482, Dmel\CG14482, Dmel_CG14482, UCRY
Chromosome Location
Chr.2R: 17572096 - 17572784 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034245
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274128.2 NP_001261057.1 1 - 2 129 66
NM_137389.4 NP_611233.1 1 - 2 129 66
GenBank mRNA BT022080.1 - 1 - 2 100 66
KX531550.1 - 1 - 2 49 66

View Full Product Details