Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Nup75
- Entrez Gene ID
-
37078
- Gene Name
- Nucleoporin 75kD
- Gene Aliases
-
CG5733, Dmel\CG5733, Dmel_CG5733, Nup 75, nup75
- Chromosome Location
-
Chr.2R: 18164694 - 18167140 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034310
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137456.3 |
NP_611300.2 |
5 - 6 |
1949 |
62 |
View Full Product Details